(1,5-Cyclooctadiene)iridium(I)-2,4-pentanedionate
Supplier: TCI America
|
Warning
|
A2981-1G
A2981-200MG
76218-772EA
578.28
CAD
76218-772
76218-774
(1,5-Cyclooctadiene)iridium(I)-2,4-pentanedionate
(1,5-Cyclooctadiene)iridium(I)-2,4-pentanedionate
Formula:
C₁₃H₁₉O₂Ir Melting Pt: 155°C |
MDL Number:
MFCD08273779 CAS Number: 12154-84-6 |
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to crystaline |
Elemental analysis (Carbon) | 37.10 to 41.00 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...