4-(Trifluoromethyl)phenylacetylene ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 4-Ethynylbenzotrifluoride, 4-Ethynyl-alpha,alpha,alpha-trifluorotoluene, 1-ethynyl-4-(trifluoromethyl)benzene

E0626-1G E0626-5G
TCE0626-1GEA 171.9 CAD
TCE0626-1G TCE0626-5G
4-(Trifluoromethyl)phenylacetylene ≥98.0% (by GC)
4-(Trifluoromethyl)phenylacetylene
Formula: C₉H₅F₃
MW: 170.134 g/mol
Boiling Pt: 60°C
Flash Pt: 20°C
Storage Temperature: Refrigerator
MDL Number: MFCD01861903
CAS Number: 705-31-7
UN: 1993
ADR: 3 II

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR