Ethyl (4-bromobenzoyl)acetate ≥98.0% (by GC)

Supplier: TCI America

E1117-1G E1117-5G
TCE1117-1GEA 260.7 CAD
TCE1117-1G TCE1117-5G
Ethyl (4-bromobenzoyl)acetate ≥98.0% (by GC)
Ethyl (4-bromobenzoyl)acetate
Formula: C₁₁H₁₁BrO₃
Boiling Pt: 124°C
MDL Number: MFCD00231243
CAS Number: 26510-95-2

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR