Sodium pivalate hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Trimethylacetic acid sodium salt hydrate, Sodium trimethylacetate hydrate, Propanoic acid, 2,2-dimethyl-, sodium salt, hydrate

S0978-25G S0978-5G
TCS0978-25GEA 270.18 CAD
TCS0978-25G TCS0978-5G
Sodium pivalate hydrate ≥98.0% (by titrimetric analysis)
Sodium pivalate hydrate
Formula: C₅H₁₁NaO₃
MW: 142.13 g/mol
MDL Number: MFCD00150782
CAS Number: 143174-36-1
EINECS: 214-678-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water 6.5 to 12.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR