3-Benzyloxy-1-propanol ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
1-Propanol, 3-(benzyloxy)-, 3-phenylmethoxy-1-propanol, Trimethylene glycol monobenzyl ether, 3-(Benzyloxy)-1-propanol, 3-(Benzyloxy)propan-1-ol
B3277-25G
B3277-5G
TCB3277-25GEA
265.44
CAD
TCB3277-25G
TCB3277-5G
3-Benzyloxy-1-propanol ≥98.0% (by GC)
3-Benzyloxy-1-propanol
Formula:
C₁₀H₁₄O₂ MW: 166.22 g/mol Boiling Pt: 120°C |
MDL Number:
MFCD00029659 CAS Number: 4799-68-2 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...