Triacetyl resveratrol ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
(E)-5-(4-Acetoxystyryl)-1,3-phenylene diacetate, 3,5,4′-Tri-O-acetylresveratrol, Acetyl-trans-resveratrol, 5-[(1E)-2-[4-(acetyloxy)phenyl]ethenyl]-1,3-Benzenediol-1,3-diacetate
T3232-1G
T3232-5G
TCT3232-1GEA
28.44
CAD
TCT3232-1G
TCT3232-5G
Triacetyl resveratrol ≥98.0% (by GC)
Triacetyl resveratrol
Formula:
C₂₀H₁₈O₆ MW: 354.35 g/mol |
MDL Number:
MFCD01546481 CAS Number: 42206-94-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 117.0 to 121.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...