Mubritinib ≥98.0% (by HPLC)

Supplier: TCI America

M3058-100MG M3058-25MG
TCM3058-100MGEA 786.84 CAD
TCM3058-100MG TCM3058-25MG
Mubritinib ≥98.0% (by HPLC)
Mubritinib
Formula: C₂₅H₂₃F₃N₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09954135
CAS Number: 366017-09-6

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 158.0 to 162.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR