7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H) ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
AMCA, 2H-1-Benzopyran-3-acetic acid, 7-Amino-4-methylcoumarin-3-acetic acid, Aminomethyl coumarin acetic acid
A2820-250MG
A2820-50MG
TCA2820-250MGEA
248.06
CAD
TCA2820-250MG
TCA2820-50MG
7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H) ≥95.0% (by titrimetric analysis)
7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H)
Formula:
C₁₂H₁₁NO₄ |
MDL Number:
MFCD00081051 CAS Number: 106562-32-7 |
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 93.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...