Regorafenib ≥98.0% (by HPLC)

Supplier: TCI America
Warning

R0142-100MG R0142-25MG
TCR0142-100MGEA 369.72 CAD
TCR0142-100MG TCR0142-25MG
Regorafenib ≥98.0% (by HPLC)
Regorafenib
Formula: C₂₁H₁₅ClF₄N₄O₃
Storage Temperature: Freezer
MDL Number: MFCD16038047
CAS Number: 755037-03-7

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 11.00 to 12.20 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR