trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl ≥98.0% (by GC)
Supplier: TCI America
P2314-25G
P2314-5G
TCP2314-25GEA
733.91
CAD
TCP2314-25G
TCP2314-5G
trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl ≥98.0% (by GC)
trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl
Formula:
C₂₁H₂₉F₃ |
MDL Number:
MFCD09838998 CAS Number: 131819-23-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 64.0 to 68.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...