tert-Butyl hydrogen malonate ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M2687-25G M2687-5G
TCM2687-25GEA 145.36 CAD
TCM2687-25G TCM2687-5G
tert-Butyl hydrogen malonate ≥97.0% (by GC, titration analysis)
tert-Butyl hydrogen malonate
Formula: C₇H₁₂O₄
Boiling Pt: 90°C
Melting Pt: 19°C
Flash Pt: 91°C
MDL Number: MFCD00191886
CAS Number: 40052-13-9

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR