Phenylsuccinic anhydride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
P2257-25G
P2257-5G
TCP2257-25GEA
1042.8
CAD
TCP2257-25G
TCP2257-5G
Phenylsuccinic anhydride ≥98.0% (by GC, titration analysis)
Phenylsuccinic anhydride
Formula:
C₁₀H₈O₃ Boiling Pt: 195°C |
MDL Number:
MFCD00192395 CAS Number: 1131-15-3 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Methanolysis Method) | min. 98.0 % |
Melting point | 52.0 to 56.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...