Phenyl methacrylate ≥97.0% (by GC) stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

P2134-25G P2134-5G
TCP2134-25GEA 314.42 CAD
TCP2134-25G TCP2134-5G
Phenyl methacrylate ≥97.0% (by GC) stabilized
Phenyl methacrylate
Formula: C₁₀H₁₀O₂
Boiling Pt: 198°C
Flash Pt: 60°C
MDL Number: MFCD00048117
CAS Number: 2177-70-0
EINECS: 218-542-3
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR