Paromomycin sulfate ≥94.0% (by HPLC)

Supplier: TCI America
Warning

P2092-1G P2092-5G
TCP2092-1GEA 86.9 CAD
TCP2092-1G TCP2092-5G
Paromomycin sulfate ≥94.0% (by HPLC)
Paromomycin sulfate
Formula: C₂₃H₄₅N₅O₁₄·H₂SO₄
MDL Number: MFCD00079278
CAS Number: 1263-89-4
EINECS: 215-031-7
Merck Index: 14,07041

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 94.0 area%
Specific rotation [a]20/D +47.0 to +55.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR