O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
T2929-25G
T2929-5G
TCT2929-25GEA
134.3
CAD
TCT2929-25G
TCT2929-5G
O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)
O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate
Formula:
C₉H₁₆F₆N₃O₃P Storage Temperature: Freezer |
MDL Number:
MFCD01863753 CAS Number: 265651-18-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...