Nefazodone hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

N1030-1G N1030-200MG
TCN1030-1GEA 149.74 CAD
TCN1030-1G TCN1030-200MG
Nefazodone hydrochloride ≥98.0% (by HPLC, titration analysis)
Nefazodone hydrochloride
Formula: C₂₅H₃₂ClN₅O₂·HCl
MDL Number: MFCD00935760
CAS Number: 82752-99-6
Merck Index: 14,06438

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 186.0 to 190.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR