N-(4-Cyanophenyl)guanidine ≥99.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

C3387-1G C3387-5G
76218-932EA 44.24 CAD
76218-932 76218-934
N-(4-Cyanophenyl)guanidine ≥99.0% (by HPLC, total nitrogen)
N-(4-Cyanophenyl)guanidine
Formula: C₈H₈N₄
MDL Number: MFCD16251323
CAS Number: 5637-42-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Melting point (Decomposition) 210.0 to 214.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR