Methyl-4-hydroxy-3,5-dimethoxybenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

M2806-25G M2806-5G
TCM2806-25GEA 249.64 CAD
TCM2806-25G TCM2806-5G
Methyl-4-hydroxy-3,5-dimethoxybenzoate ≥98.0% (by GC)
Methyl-4-hydroxy-3,5-dimethoxybenzoate
Formula: C₁₀H₁₂O₅
MDL Number: MFCD00017199
CAS Number: 884-35-5
EINECS: 429-050-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 106.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR