Imipramine hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine hydrochloride

I0971-1G I0971-5G
TCI0971-1GEA 79 CAD
TCI0971-1G TCI0971-25G TCI0971-5G
Imipramine hydrochloride ≥98.0% (by HPLC)
Imipramine hydrochloride
Formula: C₁₉H₂₄N₂·HCl
MW: 316.87 g/mol
MDL Number: MFCD00012669
CAS Number: 113-52-0
EINECS: 204-030-7
Merck Index: 14,04920

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 171.0 to 175.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR