Erbstatin analog ≥96.0% (by HPLC)

Supplier: TCI America
Warning

M2520-100MG M2520-10MG
TCM2520-100MGEA 709.42 CAD
TCM2520-100MG TCM2520-10MG
Erbstatin analog ≥96.0% (by HPLC)
Erbstatin analog
Formula: C₁₀H₁₀O₄
MDL Number: MFCD00132932
CAS Number: 63177-57-1

Order Now

Specification Test Results

Purity (HPLC) min. 96.0 area%
Melting point 180.0 to 184.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR