Dimethyl iminodiacetate hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Dimethyl 2,2'-azanediyldiacetate hydrochloride

D5385-25G D5385-5G
TCD5385-25GEA 257.54 CAD
TCD5385-25G TCD5385-5G
Dimethyl iminodiacetate hydrochloride ≥98.0% (by titrimetric analysis)
Dimethyl iminodiacetate hydrochloride
Formula: C₆H₁₁NO₄·HCl
MW: 197.62 g/mol
MDL Number: MFCD00216724
CAS Number: 39987-25-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR