Diisodecyl-4-cyclohexene-1,2-dicarboxylate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

T0603-25ML T0603-500ML
TCT0603-25MLEA 67.94 CAD
TCT0603-25ML TCT0603-500ML
Diisodecyl-4-cyclohexene-1,2-dicarboxylate ≥97.0% (by titrimetric analysis)
Diisodecyl-4-cyclohexene-1,2-dicarboxylate
Formula: C₂₈H₅₀O₄
MDL Number: MFCD00070615
CAS Number: 87826-26-4

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (Ester Value) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR