Diamond nanoparticles 3% dispersion in ethylene glycol (Amine-modified) (<10 nm)

Supplier: TCI America
Danger

N1083-25G N1083-5G
TCN1083-25GEA 722.38 CAD
TCN1083-25G TCN1083-5G
Diamond nanoparticles 3% dispersion in ethylene glycol (Amine-modified) (<10 nm)
Diamond
MDL Number: MFCD00211867
CAS Number: 7782-40-3

Order Now

Specification Test Results

Appearance Light gray to Brown to Black clear liquid to cloudy liquid
Concentration 2.5 to 3.4 %
Water max. 1.0 %
Mean particle diameter max. 10 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR