Cytidine-5'-diphosphocholine sodium salt (CDP choline) ≥98.0% (by HPLC)

Supplier: TCI America

C3445-1G C3445-5G
TCC3445-1GEA 33.18 CAD
TCC3445-1G TCC3445-5G
Cytidine-5'-diphosphocholine sodium salt (CDP choline) ≥98.0% (by HPLC)
Cytidine-5'-diphosphocholine sodium salt (CDP choline)
Formula: C₁₄H₂₆N₄NaO₁₁P₂
Melting Pt: 250°C
Storage Temperature: Freezer
MDL Number: MFCD00037486
CAS Number: 33818-15-4
EINECS: 251-689-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 % (calcd.on anh.substance)
Water max. 6.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR