Chalcone-α,β-epoxide ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D4378-1G
D4378-5G
TCD4378-1GEA
41.08
CAD
TCD4378-1G
TCD4378-5G
Chalcone-α,β-epoxide ≥98.0% (by GC)
Chalcone-alpha,beta-epoxide
Formula:
C₁₅H₁₂O₂ |
MDL Number:
MFCD00022336 CAS Number: 5411-12-1 EINECS: 226-487-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...