Camostat methanesulfate ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: Camostat Methanesulfonate, 4-(2-(2-(Dimethylamino)-2-oxoethoxy)-2-oxoethyl)phenyl 4-guanidinobenzoate methanesulfonate, Camostat Mesylate, Camostat Methanesulphonate, 4-(2-(2-(Dimethylamino)-2-oxoethoxy)-2-oxoethyl)phenyl 4-guanidinobenzoate methanesulphonate

C2977-100MG C2977-25MG
TCC2977-100MGEA 415.54 CAD
TCC2977-100MG TCC2977-25MG
Camostat methanesulfate ≥98.0% (by HPLC)
Camostat methanesulfate
Formula: C₂₀H₂₂N₄O₅·CH₃SO₃H
MW: 494.52 g/mol
MDL Number: MFCD00941410
CAS Number: 59721-29-8
Merck Index: 14,01728

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 11.10 to 11.60 %
Melting point 196.0 to 200.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR