Bisacodyl ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

B5066-25G B5066-5G
TCB5066-25GEA 374.46 CAD
TCB5066-25G TCB5066-5G
Bisacodyl ≥98.0% (by HPLC, titration analysis)
Bisacodyl
Formula: C₂₂H₁₉NO₄
MDL Number: MFCD00038039
CAS Number: 603-50-9
EINECS: 210-044-4
Merck Index: 14,01242

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 132.0 to 136.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR