9,9-Dimethylfluorene-2-boronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D5018-1G
D5018-5G
76219-776EA
63.2
CAD
76219-776
76219-778
9,9-Dimethylfluorene-2-boronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
9,9-Dimethylfluorene-2-boronic acid pinacol ester
Formula:
C₂₁H₂₅BO₂ |
MDL Number:
MFCD08704229 CAS Number: 569343-09-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 134.0 to 138.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...