9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 9,9-Dimethylfluorene-2,7-diboronic acid Bis(pinacol) ester, 2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-dimethylfluorene, 2,2'-(9,9-Dimethyl-9H-fluorene-2,7-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

B5157-1G B5157-5G
76219-508EA 71.1 CAD
76219-508 76219-510
9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester ≥98.0% (by HPLC, titration analysis)
9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester
Formula: C₂₇H₃₆B₂O₄
MW: 446.20 g/mol
MDL Number: MFCD08704230
CAS Number: 325129-69-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR