7-Bromo-2H-1,4-benzothiazin-3(4H)-one ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 7-Bromo-2H-benzo[b][1,4]thiazin-3(4H)-one

B4902-1G B4902-5G
TCB4902-1GEA 153.26 CAD
TCB4902-1G TCB4902-5G
7-Bromo-2H-1,4-benzothiazin-3(4H)-one ≥98.0% (by GC)
7-Bromo-2H-1,4-benzothiazin-3(4H)-one
Formula: C₈H₆BrNOS
MW: 244.11 g/mol
MDL Number: MFCD02660583
CAS Number: 90814-91-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 212.0 to 216.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR