6-Chloro-2-iodo-3-hydroxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C3366-1G C3366-5G
TCC3366-1GEA 124.82 CAD
TCC3366-1G TCC3366-5G
6-Chloro-2-iodo-3-hydroxypyridine ≥98.0% (by GC, titration analysis)
6-Chloro-2-iodo-3-hydroxypyridine
Formula: C₅H₃ClIO
MDL Number: MFCD07636749
CAS Number: 188057-26-3

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR