5-Bromo-2-(2-thienyl)pyridine 98,0 GC, T
Supplier: TCI America
|
Warning
|
Synonyms:
5-Bromo-2-(thiophen-2-yl)pyridine
B4885-1G
B4885-200MG
TCB4885-1GEA
447.14
CAD
TCB4885-1G
TCB4885-200MG
5-Bromo-2-(2-thienyl)pyridine 98,0 GC, T
5-Bromo-2-(2-thienyl)pyridine
Formula:
C₉H₆BrNS MW: 240.12 g/mol |
MDL Number:
MFCD09037419 CAS Number: 91891-74-6 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 79.0 to 83.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...