5-Amino-4-bromo-3-methyl-1-phenyl-1H-pyrazole ≥96.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
5-Amino-4-bromo-3-methyl-1-phenylpyrazole
A2901-1G
A2901-5G
TCA2901-1GEA
101.12
CAD
TCA2901-1G
TCA2901-5G
5-Amino-4-bromo-3-methyl-1-phenyl-1H-pyrazole ≥96.0% (by titrimetric analysis)
5-Amino-4-bromo-3-methyl-1-phenyl-1H-pyrazole
Formula:
C₁₀H₁₀BrN₃ MW: 252.12 g/mol |
MDL Number:
MFCD04039926 CAS Number: 69464-98-8 |
Specification Test Results
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 102.0 to 106.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...