5-Acetyl-2-amino-4-methylthiazole ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
A2887-1G
A2887-5G
TCA2887-1GEA
61.62
CAD
TCA2887-1G
TCA2887-5G
5-Acetyl-2-amino-4-methylthiazole ≥98.0% (by GC, titration analysis)
5-Acetyl-2-amino-4-methylthiazole
Formula:
C₆H₈N₂OS Melting Pt: 266°C |
MDL Number:
MFCD00051952 CAS Number: 30748-47-1 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...