4-Cyano-4''-n-pentyl-p-terphenyl ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4''-Pentyl-[1,1':4',1''-terphenyl]-4-carbonitrile, 4-Cyano-4''-pentyl-p-terphenyl

C2910-1G C2910-5G
TCC2910-1GEA 193.39 CAD
TCC2910-1G TCC2910-5G
4-Cyano-4''-n-pentyl-p-terphenyl ≥98.0% (by GC)
4-Cyano-4''-n-pentyl-p-terphenyl
Formula: C₂₄H₂₃N
MW: 325.45 g/mol
Melting Pt: 130°C
MDL Number: MFCD00799423
CAS Number: 54211-46-0
EINECS: 259-028-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 128 to 240 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR