4-Acetyl-4'-methoxydiphenyl ether ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-(4-(4-Methoxyphenoxy)phenyl)ethanone, 4'-(4-Methoxyphenoxy)acetophenone, 4-Acetyl-4'-methoxydiphenyl ether

M2740-1G M2740-5G
TCM2740-1GEA 113.76 CAD
TCM2740-1G TCM2740-5G
4-Acetyl-4'-methoxydiphenyl ether ≥98.0% (by GC)
4-Acetyl-4'-methoxydiphenyl ether
Formula: C₁₅H₁₄O₃
MW: 242.28 g/mol
Boiling Pt: 236°C
MDL Number: MFCD00052928
CAS Number: 54916-28-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR