4-Acetoxybenzeneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

A2818-1G A2818-200MG
TCA2818-1GEA 172.22 CAD
TCA2818-1G TCA2818-200MG
4-Acetoxybenzeneboronic acid (contains varying amounts of Anhydride)
4-Acetoxybenzeneboronic acid
Formula: C₈H₉BO₄
MDL Number: MFCD09027198
CAS Number: 177490-82-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR