4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-iodoquinazolin-4-amine

C2965-1G C2965-5G
TCC2965-1GEA 82.63 CAD
TCC2965-1G TCC2965-5G
4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline ≥98.0% (by HPLC, titration analysis)
4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline
Formula: C₂₁H₁₄ClFIN₃O
MW: 505.72 g/mol
MDL Number: MFCD09998827
CAS Number: 231278-20-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 223.0 to 227.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR