4,4'-Dichlorochalcone ≥98.0% (by GC)

Supplier: TCI America
Warning

D4846-25G D4846-5G
TCD4846-25GEA 641.48 CAD
TCD4846-25G TCD4846-5G
4,4'-Dichlorochalcone ≥98.0% (by GC)
4,4'-Dichlorochalcone
Formula: C₁₅H₁₀Cl₂O
MDL Number: MFCD00018704
CAS Number: 19672-59-4

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 156.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR