4-(n-Butoxy)phenylacetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B4742-1G
B4742-5G
TCB4742-1GEA
48.98
CAD
TCB4742-1G
TCB4742-5G
4-(n-Butoxy)phenylacetic acid ≥98.0% (by GC, titration analysis)
4-(n-Butoxy)phenylacetic acid
Formula:
C₁₂H₁₆O₃ |
MDL Number:
MFCD00016832 CAS Number: 4547-57-3 EINECS: 224-904-1 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 85.0 to 89.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...