4-(9-Carbazolyl)benzeneboronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T3452-1G T3452-5G
TCT3452-1GEA 60.04 CAD
TCT3452-1G TCT3452-5G
4-(9-Carbazolyl)benzeneboronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
4-(9-Carbazolyl)benzeneboronic acid pinacol ester
Formula: C₂₄H₂₄BNO₂
MDL Number: MFCD16294549
CAS Number: 785051-54-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 176.0 to 180.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR