3-Amino-2-methoxypyridine-5-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine, 3-Amino-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Methoxy-3-aminopyridine-5-boronic acid pinacol ester, 2-(5-Amino-6-methoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Amino-2-methoxypyridine-5-boronic acid Pinacol ester

A2993-1G A2993-200MG
TCA2993-1GEA 347.6 CAD
TCA2993-1G TCA2993-200MG
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester
Formula: C₁₂H₁₉BN₂O₃
MW: 250.10 g/mol
MDL Number: MFCD12923427
CAS Number: 893440-50-1

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 142.0 to 146.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR