3,3'-Diethylthiatricarbocyanine iodide ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Warning

D4773-1G D4773-5G
TCD4773-1GEA 131.84 CAD
TCD4773-1G TCD4773-5G
3,3'-Diethylthiatricarbocyanine iodide ≥98.0% (by total nitrogen basis)
3,3'-Diethylthiatricarbocyanine iodide
Formula: C₂₅H₂₅IN₂S₂
Melting Pt: 211°C
MDL Number: MFCD00041994
CAS Number: 3071-70-3
EINECS: 221-342-9

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR