3-(2-Thienyl)pyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

T3055-1G T3055-200MG
TCT3055-1GEA 581.44 CAD
TCT3055-1G TCT3055-200MG
3-(2-Thienyl)pyridine ≥98.0% (by GC)
3-(2-Thienyl)pyridine
Formula: C₉H₇NS
Boiling Pt: 100°C
MDL Number: MFCD01860052
CAS Number: 21298-53-3

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR