2-Chloro-1,3-dimethylimidazolium tetrafluoroborate ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
C3444-1G
C3444-5G
TCC3444-1GEA
80.58
CAD
TCC3444-1G
TCC3444-5G
2-Chloro-1,3-dimethylimidazolium tetrafluoroborate ≥98.0% (by HPLC)
2-Chloro-1,3-dimethylimidazolium tetrafluoroborate
Formula:
C₅H₁₀ClN₂·BF₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD09039291 CAS Number: 153433-26-2 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 187.0 to 191.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...