2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride ≥98.0% (by HPLC) (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

A3028-1G A3028-200MG
TCA3028-1GEA 284.4 CAD
TCA3028-1G TCA3028-200MG
2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride
Formula: C₈H₁₀BNO₄·HCl
Melting Pt: 180°C
MDL Number: MFCD02258941
CAS Number: 380430-55-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 97.0 to 108.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR