1-Boc-azetidine-3-carboxaldehyde ≥98.0% (by GC)

Supplier: TCI America

B5160-1G B5160-200MG
76219-512EA 249.64 CAD
76219-512 76219-514
1-Boc-azetidine-3-carboxaldehyde ≥98.0% (by GC)
1-Boc-azetidine-3-carboxaldehyde
Formula: C₉H₁₅NO₃
MDL Number: MFCD06656142
CAS Number: 177947-96-5

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR