1-Benzylpiperidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B4698-25ML
B4698-5ML
TCB4698-25MLEA
213.3
CAD
TCB4698-25ML
TCB4698-5ML
1-Benzylpiperidine ≥98.0% (by GC, titration analysis)
1-Benzylpiperidine
Formula:
C₁₂H₁₇N Boiling Pt: 66°C |
MDL Number:
MFCD00224901 CAS Number: 2905-56-8 EINECS: 220-809-4 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...