1,4-Bis(2-hydroxyethyl)piperazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2,2'-(Piperazine-1,4-diyl)diethanol

B4903-25G B4903-5G
TCB4903-25GEA 194.34 CAD
TCB4903-25G TCB4903-5G
1,4-Bis(2-hydroxyethyl)piperazine ≥98.0% (by GC, titration analysis)
1,4-Bis(2-hydroxyethyl)piperazine
Formula: C₈H₁₈N₂O₂
MW: 174.24 g/mol
Boiling Pt: 278°C
MDL Number: MFCD00006157
CAS Number: 122-96-3
EINECS: 204-586-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 136.0 to 140.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR