1,1'-Thiocarbonyldi-2(1H)-pyridone ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 1,1'-Thiocarbonylbis(pyridin-2(1H)-one)

T3644-1G T3644-5G
76220-696EA 58.46 CAD
76220-696 76220-698
1,1'-Thiocarbonyldi-2(1H)-pyridone ≥98.0% (by HPLC, total nitrogen)
1,1'-Thiocarbonyldi-2(1H)-pyridone
Formula: C₁₁H₈N₂O₂S
MDL Number: MFCD00075238
CAS Number: 102368-13-8

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 156.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR